Assay Details
Gene Symbol: AKR4C10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129335.5 | NP_181315.2 | 1 - 2 | 258 | 136 | |
GenBank mRNA | AK227354.1 | - | 1 - 2 | 163 | 136 | |
BT005862.1 | - | 1 - 2 | 112 | 136 | ||
DQ837655.1 | - | 1 - 2 | 112 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
