Assay Details
Gene Symbol: AT2G38390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129395.3 | NP_181373.1 | 3 - 4 | 635 | 66 | |
GenBank mRNA | AF452385.1 | - | 3 - 4 | 616 | 66 | |
AY099555.1 | - | 3 - 4 | 633 | 66 | ||
BT001238.1 | - | 3 - 4 | 588 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
