Assay Details
Gene Symbol: AT2G38730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129430.4 | NP_181407.1 | 5 - 6 | 623 | 72 | |
GenBank mRNA | AY568522.1 | - | 5 - 6 | 489 | 72 | |
BT002031.1 | - | 5 - 6 | 554 | 72 | ||
BT006565.1 | - | 5 - 6 | 489 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
