Assay Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129555.4 | NP_181526.1 | 5 - 6 | 698 | 79 | |
GenBank mRNA | AB047148.1 | - | 4 - 5 | 367 | 79 | |
AY039858.1 | - | 5 - 6 | 515 | 79 | ||
BT003020.1 | - | 4 - 5 | 367 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
