Assay Details
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202813.1 | NP_001189742.1 | 2 - 3 | 308 | 103 | |
NM_129896.4 | NP_181863.1 | 2 - 3 | 316 | 103 | ||
GenBank mRNA | AY065372.1 | - | 2 - 3 | 303 | 103 | |
AY087030.1 | - | 2 - 3 | 301 | 103 | ||
AY096479.1 | - | 2 - 3 | 235 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
