Assay Details
Gene Symbol: OASB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202816.1 | NP_001189745.1 | 2 - 3 | 394 | 96 | |
NM_129937.4 | NP_181903.1 | 1 - 2 | 561 | 96 | ||
GenBank mRNA | AK226461.1 | - | 1 - 2 | 476 | 96 | |
AY065375.1 | - | 1 - 2 | 369 | 96 | ||
AY086156.1 | - | 1 - 2 | 400 | 96 | ||
AY096681.1 | - | 1 - 2 | 245 | 96 | ||
X80377.2 | - | 257 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
