Assay Details
Gene Symbol: OASB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202816.1 | NP_001189745.1 | 6 - 7 | 960 | 76 | |
NM_129937.4 | NP_181903.1 | 5 - 6 | 1127 | 76 | ||
GenBank mRNA | AK226461.1 | - | 5 - 6 | 1042 | 76 | |
AY065375.1 | - | 5 - 6 | 935 | 76 | ||
AY086156.1 | - | 5 - 6 | 966 | 76 | ||
AY096681.1 | - | 5 - 6 | 811 | 76 | ||
X80377.2 | - | 823 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
