Assay Details
Gene Symbol: OASB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202816.1 | NP_001189745.1 | 10 - 11 | 1239 | 58 | |
NM_129937.4 | NP_181903.1 | 9 - 10 | 1406 | 58 | ||
GenBank mRNA | AK226461.1 | - | 9 - 10 | 1321 | 58 | |
AY065375.1 | - | 9 - 10 | 1214 | 58 | ||
AY086156.1 | - | 9 - 10 | 1245 | 58 | ||
AY096681.1 | - | 9 - 10 | 1090 | 58 | ||
X80377.2 | - | 1102 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
