Assay Details
Gene Symbol: AT2G43860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129948.2 | NP_181914.1 | 1 - 2 | 463 | 92 | |
GenBank mRNA | DQ056577.1 | - | 1 - 2 | 463 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
