Assay Details
Gene Symbol: AT2G43890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129951.2 | NP_181917.1 | 1 - 2 | 530 | 71 | |
GenBank mRNA | DQ056578.1 | - | 1 - 2 | 437 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
