Assay Details
Gene Symbol: AT2G43890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129951.2 | NP_181917.1 | 3 - 4 | 1054 | 103 | |
GenBank mRNA | DQ056578.1 | - | 3 - 4 | 961 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
