Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201881.2 | NP_973610.1 | 5 - 6 | 925 | 158 | |
NM_201882.3 | NP_973611.1 | 4 - 5 | 860 | 158 | ||
GenBank mRNA | AJ271597.1 | - | 5 - 6 | 925 | 158 | |
AJ294534.1 | - | 4 - 5 | 688 | 158 | ||
BT026376.1 | - | 4 - 5 | 688 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
