Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201881.2 | NP_973610.1 | 6 - 7 | 983 | 62 | |
NM_201882.3 | NP_973611.1 | 5 - 6 | 918 | 62 | ||
GenBank mRNA | AJ271597.1 | - | 6 - 7 | 983 | 62 | |
AJ294534.1 | - | 5 - 6 | 746 | 62 | ||
BT026376.1 | - | 5 - 6 | 746 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
