Assay Details
Gene Symbol: AT2G18150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127372.4 | NP_179407.1 | 2 - 3 | 560 | 175 | |
GenBank mRNA | AY081298.1 | - | 2 - 3 | 503 | 175 | |
BT002557.1 | - | 2 - 3 | 437 | 175 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
