Assay Details
Gene Symbol: AT2G18150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127372.4 | NP_179407.1 | 3 - 4 | 724 | 123 | |
GenBank mRNA | AY081298.1 | - | 3 - 4 | 667 | 123 | |
BT002557.1 | - | 3 - 4 | 601 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
