Assay Details
Gene Symbol: ACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335856.1 | NP_001323663.1 | 2 - 3 | 712 | 113 | |
NM_127924.3 | NP_179941.1 | 2 - 3 | 545 | 113 | ||
GenBank mRNA | AF361627.1 | - | 2 - 3 | 526 | 113 | |
AK226527.1 | - | 2 - 3 | 531 | 113 | ||
AY056068.1 | - | 2 - 3 | 513 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
