Assay Details
Gene Symbol: MYB100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128080.1 | NP_180095.1 | 1 - 2 | 106 | 137 | |
GenBank mRNA | AF176004.1 | - | 1 - 2 | 106 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
