Assay Details
Gene Symbol: MYB100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128080.1 | NP_180095.1 | 2 - 3 | 235 | 164 | |
GenBank mRNA | AF176004.1 | - | 2 - 3 | 235 | 164 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
