Assay Details
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202716.1 | NP_001189645.1 | 14 - 15 | 1322 | 151 | |
NM_128643.4 | NP_180648.1 | 13 - 14 | 1279 | 151 | ||
NM_201841.1 | NP_973570.1 | 12 - 13 | 1238 | 151 | ||
GenBank mRNA | AK227543.1 | - | 13 - 14 | 1238 | 151 | |
BT004616.1 | - | 13 - 14 | 1005 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
