Assay Details
Gene Symbol: ASB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198163.1 | NP_001185092.1 | 7 - 8 | 842 | 155 | |
NM_102331.3 | NP_173893.1 | 7 - 8 | 863 | 155 | ||
GenBank mRNA | AK226500.1 | - | 7 - 8 | 759 | 155 | |
AY099834.1 | - | 7 - 8 | 754 | 155 | ||
BT006303.1 | - | 7 - 8 | 732 | 155 | ||
L22585.1 | - | 7 - 8 | 748 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
