Assay Details
Gene Symbol: MYB59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125370.4 | NP_200786.1 | 2 - 3 | 462 | 152 | |
NM_180894.4 | NP_851225.1 | 1 - 2 | 539 | 152 | ||
NM_180895.3 | NP_851226.1 | 2 - 3 | 460 | 152 | ||
GenBank mRNA | AK229048.1 | - | 1 - 2 | 442 | 152 | |
AY519641.1 | - | 2 - 3 | 250 | 152 | ||
DQ075252.1 | - | 1 - 2 | 361 | 152 | ||
DQ075253.1 | - | 2 - 3 | 282 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_180894.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
