Assay Details
Gene Symbol: BGLU34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198236.1 | NP_001185165.1 | 6 - 7 | 589 | 167 | |
NM_103653.3 | NP_175191.2 | 6 - 7 | 590 | 167 | ||
GenBank mRNA | BT000471.1 | - | 6 - 7 | 589 | 167 | |
BT002202.1 | - | 6 - 7 | 550 | 167 | ||
BT002458.1 | - | 6 - 7 | 586 | 167 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
