Assay Details
Gene Symbol: XRCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161167.1 | NP_001154639.1 | 2 - 3 | 371 | 146 | |
NM_113211.2 | NP_188951.2 | 2 - 3 | 371 | 146 | ||
GenBank mRNA | AF233528.2 | - | 2 - 3 | 371 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
