Assay Details
Gene Symbol: Prx37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116947.3 | NP_192617.1 | 1 - 2 | 347 | 146 | |
GenBank mRNA | AF452387.1 | - | 1 - 2 | 222 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
