Assay Details
Gene Symbol: MYB33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085068.2 | NP_001078537.1 | 2 - 3 | 723 | 169 | |
NM_001342848.1 | NP_001330997.1 | 1 - 2 | 999 | 169 | ||
NM_001342849.1 | NP_001330996.1 | 2 - 3 | 900 | 169 | ||
NM_120692.3 | NP_196228.1 | 3 - 4 | 563 | 169 | ||
NM_180448.2 | NP_850779.1 | 3 - 4 | 642 | 169 | ||
GenBank mRNA | AF411969.1 | - | 3 - 4 | 468 | 169 | |
AK118937.1 | - | 3 - 4 | 563 | 169 | ||
AY519616.1 | - | 1 - 2 | 340 | 169 | ||
BT006049.1 | - | 1 - 2 | 340 | 169 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
