Assay Details
Gene Symbol: MYB119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125275.2 | NP_568891.1 | 2 - 3 | 469 | 164 | |
GenBank mRNA | AF371978.1 | - | 2 - 3 | 469 | 164 | |
DQ447091.1 | - | 2 - 3 | 469 | 164 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
