Assay Details
Gene Symbol: MYB59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125370.4 | NP_200786.1 | 1 - 2 | 333 | 150 | |
GenBank mRNA | AY519641.1 | - | 1 - 2 | 121 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
