Assay Details
Gene Symbol: UGT74F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129946.3 | NP_181912.1 | 1 - 2 | 682 | 153 | |
GenBank mRNA | KJ138854.1 | - | 1 - 2 | 621 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
