Assay Details
Gene Symbol: AT1G32630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333015.1 | NP_001322452.1 | 2 | 645 | 147 | |
NM_102997.4 | NP_174540.1 | 2 | 674 | 147 | ||
GenBank mRNA | AK220669.1 | - | 2 | 669 | 147 |
Gene Symbol: MYC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102998.4 | NP_174541.1 | 1 | 3006 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
