Assay Details
Gene Symbol: AT3G11280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111963.3 | NP_187737.1 | 3 | 1175 | 62 | |
NM_180227.4 | NP_850558.1 | 2 | 1365 | 62 | ||
GenBank mRNA | AY056180.1 | - | 2 | 903 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
