Assay Details
Gene Symbol: AT3G46430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114510.3 | NP_190227.1 | 1 | 211 | 73 | |
GenBank mRNA | AK117680.1 | - | 1 | 152 | 73 | |
AY064154.1 | - | 1 | 131 | 73 | ||
AY097410.1 | - | 1 | 62 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
