Assay Details
Gene Symbol: BGLU8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340172.1 | NP_001327941.1 | 9 - 10 | 1182 | 68 | |
NM_001340174.1 | NP_001327939.1 | 9 - 10 | 1140 | 68 | ||
NM_001340175.1 | NP_001327940.1 | 8 - 9 | 1212 | 68 | ||
NM_001340176.1 | NP_001327937.1 | 8 - 9 | 1158 | 68 | ||
NM_116140.4 | NP_191834.3 | 9 - 10 | 1110 | 68 | ||
GenBank mRNA | AK175256.1 | - | 9 - 10 | 1059 | 68 | |
AK176786.1 | - | 9 - 10 | 1058 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
