Assay Details
Gene Symbol: LLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203929.2 | NP_001190858.1 | 3 | 539 | 71 | |
NM_001341909.1 | NP_001328710.1 | 2 | 629 | 71 | ||
NM_118968.2 | NP_194557.2 | 1 | 759 | 71 | ||
GenBank mRNA | BT010922.1 | - | 1 | 759 | 71 | |
BT011635.1 | - | 1 | 361 | 71 |
Gene Symbol: AT4G28290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341910.1 | NP_001320080.1 | 3 | 430 | 71 | |
NM_118969.4 | NP_194558.1 | 2 | 1065 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
