Assay Details
Gene Symbol: EDA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336584.1 | NP_001324434.1 | 2 - 3 | 461 | 107 | |
NM_129154.4 | NP_181139.2 | 2 - 3 | 437 | 107 | ||
GenBank mRNA | AK117562.1 | - | 2 - 3 | 416 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
