Assay Details
Gene Symbol: AT2G38380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129394.3 | NP_181372.1 | 3 - 4 | 740 | 120 | |
GenBank mRNA | AF452388.1 | - | 3 - 4 | 607 | 120 | |
AK230233.1 | - | 1399 | 120 | |||
AY035033.1 | - | 3 - 4 | 645 | 120 | ||
AY059106.1 | - | 3 - 4 | 586 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
