Assay Details
Gene Symbol: MYB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121683.4 | NP_197179.2 | 3 - 4 | 1217 | 148 | |
NM_203063.2 | NP_974792.1 | 2 - 3 | 1471 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
