Assay Details
Gene Symbol: AT3G06778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084648.3 | NP_001078117.1 | 2 - 3 | 305 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT3G06778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084648.3 | NP_001078117.1 | 2 - 3 | 305 | 130 |
Set Membership: |
![]() ![]() ![]() ![]() |