Assay Details
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034747.2 | NP_001029919.1 | 1 - 2 | 150 | 88 | |
GenBank mRNA | BC103269.1 | - | 1 - 2 | 151 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103269.1, NM_001034747.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
