Assay Details
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038565.2 | NP_001033654.1 | 1 - 2 | 29 | 67 | |
GenBank mRNA | BC109848.1 | - | 1 - 2 | 30 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109848.1, NM_001038565.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
