Assay Details
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034596.2 | NP_001029768.1 | 5 - 6 | 660 | 71 | |
GenBank mRNA | BC102554.1 | - | 5 - 6 | 661 | 71 | |
BC151435.1 | - | 5 - 6 | 660 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
