Assay Details
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174310.3 | NP_776735.2 | 4 - 5 | 302 | 82 | |
GenBank mRNA | AF257235.1 | - | 4 - 5 | 301 | 82 | |
BC111272.1 | - | 4 - 5 | 302 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
