Assay Details
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046583.2 | NP_001040048.1 | 1 | 247 | 77 | |
GenBank mRNA | BC111673.1 | - | 1 | 248 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
