Assay Details
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075758.2 | NP_001069226.1 | 2 - 3 | 1394 | 105 | |
GenBank mRNA | BC120078.1 | - | 2 - 3 | 1395 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
