Assay Details
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033628.1 | NP_001028800.1 | 1 - 2 | 205 | 129 | |
GenBank mRNA | BC103436.1 | - | 1 - 2 | 205 | 129 | |
BT029815.1 | - | 1 - 2 | 197 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103436.1, NM_001033628.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
