Assay Details
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001141.1 | NP_001001141.1 | 1 - 2 | 72 | 91 | |
GenBank mRNA | L26101.2 | - | 1 - 2 | 72 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001001141.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
