Assay Details
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001141.1 | NP_001001141.1 | 3 - 4 | 409 | 116 | |
GenBank mRNA | BC147983.1 | - | 2 - 3 | 334 | 116 | |
L26101.2 | - | 3 - 4 | 409 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
