Assay Details
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002888.1 | NP_001002888.1 | 4 - 5 | 741 | 113 | |
GenBank mRNA | AY427789.1 | - | 4 - 5 | 741 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
