Assay Details
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008669.1 | NP_001008669.1 | 1 - 2 | 59 | 72 | |
GenBank mRNA | AY821679.1 | - | 1 - 2 | 59 | 72 | |
BC102375.1 | - | 1 - 2 | 141 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102375.1, NM_001008669.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
