Assay Details
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013003.1 | NP_001013021.1 | 1 - 2 | 44 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001013003.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
