Assay Details
Gene Symbol: CXCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014859.1 | NP_001014859.1 | 1 - 2 | 89 | 67 | |
GenBank mRNA | BT020675.1 | - | 1 - 2 | 89 | 67 | |
JC209416.1 | - | 1 - 2 | 89 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001014859.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
